missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium oxalate, 0.1N Standardized Solution

Manufacturer:  Alfa Aesar™ 47123AK

 View more versions of this product

Catalog No. AA47123AK



Specifications

Specifications

Sodium oxalate
62-76-0
Na2C2O4
sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
C(=O)(C(=O)[O-])[O-].[Na+].[Na+]
133.998
134
250mL
0.1N Standardized Solution
C2Na2O4
MFCD00012465
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
disodium;oxalate
6125
Liquid

Chemical Identifiers

62-76-0
133.998
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
6125
C(=O)(C(=O)[O-])[O-].[Na+].[Na+]
C2Na2O4
MFCD00012465
sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
disodium;oxalate
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents