Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium oxalate, 0.1N Standardized Solution

Chemical Identifiers

Chemical Name or Material Sodium oxalate
CAS 62-76-0
Name Note 0.1N Standardized Solution
Linear Formula Na2C2O4
Formula Weight 134
MDL Number MFCD00012465
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47123AK

  SDS
alfa aesar™
47123AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA47123K2

  SDS
alfa aesar™
47123K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 134
Chemical Name or Material Sodium oxalate
TSCA TSCA
Quantity 250mL
Name Note 0.1N Standardized Solution
Physical Form Liquid
Linear Formula Na2C2O4
MDL Number MFCD00012465
CAS 62-76-0