Please login to your online account to display your discounted pricing

Sodium Oxalate, 0.100 Molar (M/10) (0.200 Normal), Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR750000020F

 View more versions of this product

Catalog No. R750000020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 20L
Solvent or Matrix Aqueous Solution
Chemical Name or Material Sodium Oxalate
CAS 7732-18-5
Assay Percent Range 0.1000 ± 0.0001 M at 20°C
Name Note 0.100 Molar (M/10) (0.200 Normal)
Packaging Cubitainer
CAS 62-76-0
Solubility Solubility in Water: Infinite
Odor Odorless

Specifications

Chemical Name or Material Sodium Oxalate
Name Note 0.100 Molar (M/10) (0.200 Normal)
CAS 7732-18-5
CAS 62-76-0
Physical Form Liquid
Assay Percent Range 0.1000 ± 0.0001 M at 20°C