Please login to your online account to display your discounted pricing

Sodium Oxalate, 0.100 Molar (M/10) (0.200 Normal), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Oxalate
Name Note 0.100 Molar (M/10) (0.200 Normal)
CAS 7732-18-5
CAS 62-76-0
Physical Form Liquid
Assay Percent Range 0.1000 ± 0.0001 M at 20°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R75000001A

 
ricca chemical company
RIR75000001A
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R750000020F

 
ricca chemical company
RIR750000020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R75000004A

 
ricca chemical company
RIR75000004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 1L
Chemical Name or Material Sodium Oxalate
Assay Percent Range 0.1000 ± 0.0001 M at 20°C
Name Note 0.100 Molar (M/10) (0.200 Normal)
CAS 62-76-0
Solvent or Matrix Aqueous Solution
Solubility Solubility in Water: Infinite
Packaging Poly natural
Odor Odorless
CAS 7732-18-5