Please login to your online account to display your discounted pricing

Sodium Oxalate, 0.0250 Molar (M/40), 0.0500 Normal (N/20), Ricca Chemical

$53.18 - $86.58

Chemical Identifiers

Chemical Name or Material Sodium Oxalate
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 62-76-0
CAS Min % 0.33
CAS Max % 0.34
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7499-516

 
ricca chemical company
RI7499.516
Poly natural 500mL Each for $53.18

7499-532

 
ricca chemical company
RI7499.532
Poly natural 1L Each for $86.58
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 500mL
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material Sodium Oxalate
CAS 7732-18-5
Concentration or Composition (by Analyte or Components) 0.0250 Molar (M/40), 0.0500 Normal (N/20)
Shelf Life 18 months
Packaging Poly natural
CAS 62-76-0
CAS Min % 0.33
CAS Max % 0.34
Safety and Handling

18 months