missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium orthosilicate, (mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution)

Manufacturer:  Alfa Aesar™ 17568A1

 View more versions of this product

Catalog No. AA17568A1



Specifications

Specifications

Sodium orthosilicate
13472-30-5
MFCD00014252
sodium orthosilicate, tetrasodium orthosilicate, tetrasodium silicate, silicic acid h4sio4 , sodium salt 1:4, silicic acid h4sio4 , tetrasodium salt, sodium silicate, ortho, teu2jic5pa, unii-teu2jic5pa, dryorth, sodium silicate 4-
[O-][Si]([O-])([O-])[O-].[Na+].[Na+].[Na+].[Na+]
184.04
Crystalline
White
Mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution
Na4O4Si
UN1759
POWFTOSLLWLEBN-UHFFFAOYSA-N
tetrasodium;silicate
26051
Odorless
1kg

Chemical Identifiers

13472-30-5
184.04
POWFTOSLLWLEBN-UHFFFAOYSA-N
26051
[O-][Si]([O-])([O-])[O-].[Na+].[Na+].[Na+].[Na+]
Na4O4Si
MFCD00014252
sodium orthosilicate, tetrasodium orthosilicate, tetrasodium silicate, silicic acid h4sio4 , sodium salt 1:4, silicic acid h4sio4 , tetrasodium salt, sodium silicate, ortho, teu2jic5pa, unii-teu2jic5pa, dryorth, sodium silicate 4-
tetrasodium;silicate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

TSCA : TSCA

SDS Documents