Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium, Oil based standard solution, Specpure™, Na 1000μg/g

Chemical Identifiers

Chemical Name or Material Sodium Oil based standard solution
Name Note Na 1000μg/g
Purity Grade Specpure™
MDL Number MFCD00085307
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1421218

  SDS
alfa aesar™
1421218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Chemical Name or Material Sodium Oil based standard solution
MDL Number MFCD00085307
Quantity 50g
Purity Grade Specpure™
Name Note Na 1000μg/g