Please login to your online account to display your discounted pricing

Sodium Nitroprusside, Dihydrate, ACS Grade, 99.0 to 102.0%, LabChem™

$141.20 - $141.20

Chemical Identifiers

Chemical Name or Material Sodium Nitroprusside
CAS 13755-38-9
CAS Max % 100
Name Note Dihydrate
Purity Grade ACS Grade
Linear Formula Na2Fe(CN)5(NO)·2H2O
Formula Weight 297.25
UN Number UN1588
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC246909

 
lab chem
LC246909
100g Each for $141.20
Description & Specifications

Specifications

Formula Weight 297.25
Physical Form Crystals
CAS 13755-38-9
Color Dark brown
CAS Max % 100
Density 1.72g/cm3
Chemical Name or Material Sodium Nitroprusside
Identification Passes Test
Name Note Dihydrate
Packaging Amber Glass
Purity Grade ACS Grade
Assay 99.0 to 102.0%
Solubility Soluble in water
Specific Gravity 1.72g/cm3
Storage Information Room Temperature
Linear Formula Na2Fe(CN)5(NO)·2H2O
UN Number UN1588
Decomposition Information Hydrogen cyanide; Carbon monoxide; Carbon dioxide
Quantity 100g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
Toxic if swallowed.
Health Hazard 3 GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Safety and Handling

Danger

GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Toxic if swallowed.