Please login to your online account to display your discounted pricing

Sodium Nitroferricyanide, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$228.16 - $494.19

Chemical Identifiers

Chemical Name or Material Sodium Nitroferricyanide
CAS 13755-38-9
Name Note Dihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na2Fe(CN)5NO·2H2O
Formula Weight 297.95
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004280

 
avantor performance
379201
Poly Bottle 500g Each for $494.19

02004281

 
avantor performance
379204
Poly Bottle 125g Each for $228.16
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 297.95
Molecular Formula Na2Fe(CN)5NO·2H2O
Chemical Name or Material Sodium Nitroferricyanide
CAS 13755-38-9
Quantity 500g
Packaging Poly Bottle
Name Note Dihydrate