Please login to your online account to display your discounted pricing

Sodium Nitroferricyanide, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 125g
Packaging Poly Bottle

Manufacturer: avantor performance  379204

 View more versions of this product

Catalog No. 02004281

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 297.95
Physical Form Crystals
Purity Grade Baker Analyzed™ ACS Reagent
CAS 13755-38-9
Molecular Formula Na2Fe(CN)5NO·2H2O
Packaging Poly Bottle
Quantity 125g
Chemical Name or Material Sodium Nitroferricyanide
Name Note Dihydrate

Specifications

Chemical Name or Material Sodium Nitroferricyanide
CAS 13755-38-9
Name Note Dihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na2Fe(CN)5NO·2H2O
Formula Weight 297.95