missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Nitrite ACS AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7632-00-0
Physical Form Granular
Molecular Formula NaNO2
Formula Weight 69
Chemical Name or Material Sodium Nitrite
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK7824212 Avantor Performance
782406
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK7824500 Avantor Performance
782404
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7632-00-0
NaNO2
Sodium Nitrite
Granular
69
ACS Grade

Specifications

7632-00-0
NaNO2
Poly
Sodium Nitrite
Granular
69
2.5kg
ACS Grade