missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium nitrite, ACS, 97% min, Alfa Aesar™

Manufacturer:  Alfa Aesar 1424422*

 View more versions of this product

Catalog No. AA1424422



Specifications

Specifications

Sodium nitrite
7632-00-0
NNaO2
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
N(=O)[O-].[Na+]
68.995
CHEBI:78870
Granular
100g
For ACS analysis; Decomposes above 320°
97% min
MFCD00011118
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium;nitrite
23668193
69
2.17g/mL

Chemical Identifiers

7632-00-0
68.995
LPXPTNMVRIOKMN-UHFFFAOYSA-M
23668193
sodium;nitrite
NNaO2
MFCD00011118
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
CHEBI:78870
N(=O)[O-].[Na+]
Safety and Handling

Safety and Handling

EINECSNumber : 231-555-9

Documents