Please login to your online account to display your discounted pricing

Sodium Nitrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$321.57 - $1,143.63

Chemical Identifiers

Chemical Name or Material Sodium Nitrate
CAS 7631-99-4
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula NaNO3
Formula Weight 84.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004179

 
avantor performance
377005
Poly Bottle 2.5kg Each for $1,143.63

02004180

 
avantor performance
377001
Poly Bottle 500g Each for $321.57
Description & Specifications

Specifications

Chemical Name or Material Sodium Nitrate
CAS 7631-99-4
Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Molecular Formula NaNO3
Formula Weight 84.99
Packaging Poly Bottle
Specific Gravity 1L = 2.26kg
Quantity 2.5kg