Please login to your online account to display your discounted pricing

Sodium Nitrate, ACS Grade, ≥ 99.0%, LabChem™

$107.71 - $107.71

Chemical Identifiers

Chemical Name or Material Sodium Nitrate
CAS 7631-99-4
CAS Max % 100
Purity Grade ACS Grade
Linear Formula NaNO3
Formula Weight 84.99
UN Number UN1498
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC246501

 
lab chem
LC246501
500g Each for $107.71
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 84.99
Packaging Poly Bottle
Color White
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use sand, extinguishing powder to extinguish.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Boiling Point 380°C
Identification Passes Test
Purity Grade ACS Grade
Quantity 500g
UN Number UN1498
Solubility Soluble in water
Specific Gravity 2.261g/cm3
Decomposition Information Nitrogen oxides; Oxygen
Melting Point 306°C
CAS Max % 100
CAS 7631-99-4
Storage Information Room Temperature
Linear Formula NaNO3
Density 2.261g/cm3
Health Hazard 2 GHS H Statement
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause cancer (oral).
May cause damage to organs (blood, heart, liver) through prolonged or repeated exposure (oral).
Physical Form Crystals
Health Hazard 1 Danger
Chemical Name or Material Sodium Nitrate
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use sand, extinguishing powder to extinguish.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause cancer (oral).
May cause damage to organs (blood, heart, liver) through prolonged or repeated exposure (oral).