Learn More
Sodium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01087306
Description
Sodium nitrate is used in the production of fertilizers, nitric acid, pyrotechnics, smoke-bombs, glass and pottery enamels. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent. Further it finds use as food preservative and as a solid rocket propellant. It is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium nitrate | |
7631-99-4 | |
5 g | |
NNaO3 | |
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1 | |
VWDWKYIASSYTQR-UHFFFAOYSA-N | |
sodium nitrate | |
24268 | |
84.99 | |
Hygroscopic | |
2.26 g/cm3 at 20°C |
308°C | |
Powder | |
99.999% (metals basis) | |
MFCD00011119 | |
Freely soluble in water,ammonia and hydrazine; soluble in alcohol; slightly soluble in pyridine. | |
[Na+].[O-][N+]([O-])=O | |
84.99 | |
CHEBI:63005 | |
Odorless | |
White crystals |
Chemical Identifiers
7631-99-4 | |
84.99 | |
VWDWKYIASSYTQR-UHFFFAOYSA-N | |
24268 | |
sodium nitrate |
NNaO3 | |
MFCD00011119 | |
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1 | |
CHEBI:63005 | |
[Na+].[O-][N+]([O-])=O |
Safety and Handling
P210-P220-P221-P260-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
H272-H319-H373
DOTInformation : DOT Class: 5.1, Packaging Group: III
EINECSNumber : 231-554-3
RTECSNumber : WC5600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only