missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium nitrate, 99.0% min., ACS, Alfa Aesar™

Manufacturer:  Alfa Aesar 14493A7

 View more versions of this product

Catalog No. AA14493A7

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Sodium nitrate
7631-99-4
NNaO3
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
[N+](=O)([O-])[O-].[Na+]
84.994
CHEBI:63005
Crystalline
5kg
For ACS analysis
99.0% min.
MFCD00011119
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium;nitrate
24268
84.99
2.26

Chemical Identifiers

7631-99-4
84.994
VWDWKYIASSYTQR-UHFFFAOYSA-N
24268
sodium;nitrate
NNaO3
MFCD00011119
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
CHEBI:63005
[N+](=O)([O-])[O-].[Na+]
Safety and Handling

Safety and Handling

EINECSNumber : 231-554-3

Documents