Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium niobium oxide, Puratronic™, 99.997% (metals basis)

Chemical Identifiers

Chemical Name or Material Sodium niobium oxide
CAS 12034-09-2
Purity Grade Puratronic™
Formula Weight 163.89
MDL Number MFCD00014230
Synonym Sodium niobate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1087206

  SDS
alfa aesar™
1087206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1087214

  SDS
alfa aesar™
1087214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 163.89
Odor Odorless
Purity Grade Puratronic™
Assay Percent Notes (metals basis)
CAS 12034-09-2
Quantity 5g
Chemical Name or Material Sodium niobium oxide
Melting Point 1422°C
Color White to off-white
TSCA TSCA
Physical Form Powder
Synonym Sodium niobate
Assay 99.997%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00014230
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.