Learn More
Sodium nicotinate, 98%, Thermo Scientific Chemicals
$192.56 - $712.41
Chemical Identifiers
CAS | 54-86-4 |
---|---|
Molecular Formula | C6H4NNaO2 |
Molecular Weight (g/mol) | 145.093 |
MDL Number | MFCD00064352 |
InChI Key | KFLRWGSAMLBHBV-UHFFFAOYSA-M |
Synonym | sodium nicotinate, nicotinic acid sodium salt, sodium pyridine-3-carboxylate, natriumnicotinat german, unii-3zx78b4q2l, natriumnicotinat, 3-pyridinecarboxylic acid, sodium salt, nicotinic acid, sodium salt, 3-pyridinecarboxylic acid, sodium salt 1:1, nicotinic acid sodium |
PubChem CID | 23687810 |
IUPAC Name | sodium;pyridine-3-carboxylate |
SMILES | C1=CC(=CN=C1)C(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2222422
|
Thermo Scientific Chemicals
B2222422 |
100 g |
Each for $192.56
|
|
|||||
AAB2222436
|
Thermo Scientific Chemicals
B2222436 |
500 g |
Each for $712.41
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
54-86-4 | |
145.093 | |
KFLRWGSAMLBHBV-UHFFFAOYSA-M | |
23687810 | |
C1=CC(=CN=C1)C(=O)[O-].[Na+] |
C6H4NNaO2 | |
MFCD00064352 | |
sodium nicotinate, nicotinic acid sodium salt, sodium pyridine-3-carboxylate, natriumnicotinat german, unii-3zx78b4q2l, natriumnicotinat, 3-pyridinecarboxylic acid, sodium salt, nicotinic acid, sodium salt, 3-pyridinecarboxylic acid, sodium salt 1:1, nicotinic acid sodium | |
sodium;pyridine-3-carboxylate |
Specifications
54-86-4 | |
C6H4NNaO2 | |
100 g | |
KFLRWGSAMLBHBV-UHFFFAOYSA-M | |
sodium;pyridine-3-carboxylate | |
23687810 | |
98% |
>300°C | |
MFCD00064352 | |
sodium nicotinate, nicotinic acid sodium salt, sodium pyridine-3-carboxylate, natriumnicotinat german, unii-3zx78b4q2l, natriumnicotinat, 3-pyridinecarboxylic acid, sodium salt, nicotinic acid, sodium salt, 3-pyridinecarboxylic acid, sodium salt 1:1, nicotinic acid sodium | |
C1=CC(=CN=C1)C(=O)[O-].[Na+] | |
145.093 | |
145.1 | |
Sodium nicotinate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-215-1
RTECSNumber : QT2060000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only