missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium nicotinate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2222436

 View more versions of this product

Catalog No. AAB2222436



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium nicotinate
>300°C
MFCD00064352
sodium nicotinate, nicotinic acid sodium salt, sodium pyridine-3-carboxylate, natriumnicotinat german, unii-3zx78b4q2l, natriumnicotinat, 3-pyridinecarboxylic acid, sodium salt, nicotinic acid, sodium salt, 3-pyridinecarboxylic acid, sodium salt 1:1, nicotinic acid sodium
C1=CC(=CN=C1)C(=O)[O-].[Na+]
145.093
145.1
54-86-4
C6H4NNaO2
500 g
KFLRWGSAMLBHBV-UHFFFAOYSA-M
sodium;pyridine-3-carboxylate
23687810
98%

Chemical Identifiers

54-86-4
145.093
KFLRWGSAMLBHBV-UHFFFAOYSA-M
23687810
C1=CC(=CN=C1)C(=O)[O-].[Na+]
C6H4NNaO2
MFCD00064352
sodium nicotinate, nicotinic acid sodium salt, sodium pyridine-3-carboxylate, natriumnicotinat german, unii-3zx78b4q2l, natriumnicotinat, 3-pyridinecarboxylic acid, sodium salt, nicotinic acid, sodium salt, 3-pyridinecarboxylic acid, sodium salt 1:1, nicotinic acid sodium
sodium;pyridine-3-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-215-1

RTECSNumber : QT2060000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only