missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium n-Octanoate, TCI America™

Manufacturer:  TCI America O003425G

 View more versions of this product

Catalog No. O003425G



Specifications

Specifications

Sodium n-Octanoate
25g
C8H15NaO2
sodium octanoate, sodium caprylate, octanoic acid, sodium salt, sodium n-octanoate, sodium octoate, caprylic acid sodium salt, natrium octanoat, octanoic acid sodium salt, unii-9xtm81vk2b, 9xtm81vk2b
CCCCCCCC(=O)[O-].[Na+]
166.20
166.20
≥99.0% (T)
White-Yellow
1984-06-1
MFCD00058511
BYKRNSHANADUFY-UHFFFAOYSA-M
sodium;octanoate
23664772
Crystal-Powder at 20°C

Chemical Identifiers

1984-06-1
166.20
BYKRNSHANADUFY-UHFFFAOYSA-M
23664772
CCCCCCCC(=O)[O-].[Na+]
C8H15NaO2
MFCD00058511
sodium octanoate, sodium caprylate, octanoic acid, sodium salt, sodium n-octanoate, sodium octoate, caprylic acid sodium salt, natrium octanoat, octanoic acid sodium salt, unii-9xtm81vk2b, 9xtm81vk2b
sodium;octanoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0611&(7)-0973

RTECSNumber : RH0787000

TSCA : Yes

Documents