Learn More
Sodium n-octadecyl sulfate, 98% (dry wt.), water <7%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1504203
Description
Sodium n-octadecyl sulfate is used in the formulation of hair care products including shampoo, skin cleansers, and other skin care products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium n-octadecyl sulfate is used in the formulation of hair care products including shampoo, skin cleansers, and other skin care products.
Solubility
Soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Sodium n-octadecyl sulfate | |
176°C to 180°C | |
C18H37NaO4S | |
MFCD00007469 | |
3640871 | |
Soluble in water. | |
CCCCCCCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+] | |
372.54 | |
372.55 |
1120-04-3 | |
(dry wt.), water <7% | |
CH3(CH2)17OSO3Na | |
1 g | |
sodium stearyl sulfate, octadecyl sulfate sodium salt, sodium octadecyl sulfate, octadecyl sodium sulfate, sulfuric acid, monooctadecyl ester, sodium salt, sodium octadecyl sulphate, sodium monostearyl sulfate, unii-t4eww0y4ej, sodium n-octadecyl sulfate, sodium monooctadecyl sulfate | |
NWZBFJYXRGSRGD-UHFFFAOYSA-M | |
sodium;octadecyl sulfate | |
23673604 | |
98% |
Chemical Identifiers
1120-04-3 | |
372.54 | |
NWZBFJYXRGSRGD-UHFFFAOYSA-M | |
23673604 | |
CCCCCCCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+] |
C18H37NaO4S | |
MFCD00007469 | |
sodium stearyl sulfate, octadecyl sulfate sodium salt, sodium octadecyl sulfate, octadecyl sodium sulfate, sulfuric acid, monooctadecyl ester, sodium salt, sodium octadecyl sulphate, sodium monostearyl sulfate, unii-t4eww0y4ej, sodium n-octadecyl sulfate, sodium monooctadecyl sulfate | |
sodium;octadecyl sulfate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-295-0
RTECSNumber : WT1170000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only