missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium n-nonyl sulfate, 99%

$54.89 - $178.89

Chemical Identifiers

CAS 1072-15-7
Molecular Formula C9H19NaO4S
Molecular Weight (g/mol) 246.297
MDL Number MFCD00024893
InChI Key FTWCSAMTIKSPAT-UHFFFAOYSA-M
Synonym dsstox_cid_27487
PubChem CID 23677967
IUPAC Name sodium;nonyl sulfate
SMILES CCCCCCCCCOS(=O)(=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0641803 View Documents Alfa Aesar™
L0641803
1g Each for $54.89
 
AAL0641806 View Documents Alfa Aesar™
L0641806
5g Each for $178.89
 
Specifications

Chemical Identifiers

1072-15-7
246.297
FTWCSAMTIKSPAT-UHFFFAOYSA-M
23677967
CCCCCCCCCOS(=O)(=O)[O-].[Na+]
C9H19NaO4S
MFCD00024893
dsstox_cid_27487
sodium;nonyl sulfate

Specifications

1g
C9H19NaO4S
3748751
FTWCSAMTIKSPAT-UHFFFAOYSA-M
sodium;nonyl sulfate
23677967
99%
1072-15-7
MFCD00024893
dsstox_cid_27487
CCCCCCCCCOS(=O)(=O)[O-].[Na+]
246.297
246.3
Sodium n-nonyl sulfate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : WT1165000

TSCA : TSCA