Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium n-hexadecyl sulfate, 99%

$63.43 - $235.05

Chemical Identifiers

Chemical Name or Material Sodium n-hexadecyl sulfate
CAS 1120-01-0
Molecular Formula CH3(CH2)15OSO3Na
Formula Weight 344.49
MDL Number MFCD00047766
Beilstein 3578850
Synonym n-Hexadecyl sulfate sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0640206

  SDS
alfa aesar™
L0640206
5g Each for $235.05

AAL0640203

  SDS
alfa aesar™
L0640203
1g Each for $63.43
Description & Specifications

Specifications

Formula Weight 344.49
MDL Number MFCD00047766
Molecular Formula CH3(CH2)15OSO3Na
Assay 99%
Chemical Name or Material Sodium n-hexadecyl sulfate
CAS 1120-01-0
Beilstein 3578850
RTECS Number MM0225700
TSCA TSCA
Quantity 5g
Melting Point 187° to 192°C
Synonym n-Hexadecyl sulfate sodium salt

Applications
Sodium n-hexadecyl sulfate is used as an anionic surfactant.

Solubility
Soluble in water.

Notes
Incompatible with bases and strong oxidizing agents.