Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium n-heptyl sulfate, 99% (dry wt.), water <2%

$42.19 - $564.92

Chemical Identifiers

Chemical Name or Material Sodium n-heptyl sulfate
CAS 18981-98-1
Molecular Formula C7H15NaO4S
Linear Formula CH3(CH2)6OSO3Na
Formula Weight 218.24
MDL Number MFCD00051524
Beilstein 6018116
Synonym n-Heptyl sulfate sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2210403

 
alfa aesar™
B2210403
1g Each for $42.19

AAB2210414

 
alfa aesar™
B2210414
25g Each for $564.92

AAB2210406

 
alfa aesar™
B2210406
5g Each for $145.31
Description & Specifications

Specifications

Formula Weight 218.24
Chemical Name or Material Sodium n-heptyl sulfate
TSCA TSCA
Assay 99%
Assay Percent Notes (dry wt.), water <2%
Molecular Formula C7H15NaO4S
Linear Formula CH3(CH2)6OSO3Na
MDL Number MFCD00051524
CAS 18981-98-1
Beilstein 6018116
Synonym n-Heptyl sulfate sodium salt
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 199° to 202°C (dec.)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.