Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium n-heptafluorobutyrate, 98%

$37.36 - $342.04

Chemical Identifiers

Chemical Name or Material Sodium n-heptafluorobutyrate
CAS 2218-54-4
Molecular Formula C4F7NaO2
Linear Formula CF3CF2CF2CO2Na
Formula Weight 236.02
MDL Number MFCD00066168
Synonym n-Heptafluorobutyric acid sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1685406

  SDS
alfa aesar™
L1685406
5g Each for $37.36

AAL1685414

  SDS
alfa aesar™
L1685414
25g Each for $118.58

AAL1685422

  SDS
alfa aesar™
L1685422
100g Each for $342.04
Description & Specifications

Specifications

Formula Weight 236.02
Chemical Name or Material Sodium n-heptafluorobutyrate
Assay 98%
CAS 2218-54-4
TSCA TSCA
Molecular Formula C4F7NaO2
Linear Formula CF3CF2CF2CO2Na
MDL Number MFCD00066168
Synonym n-Heptafluorobutyric acid sodium salt
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 248°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.