missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium n-dodecyl sulfate (SDS), 20% aq. soln.

Manufacturer:  Alfa Aesar™ J63394AK

 View more versions of this product

Catalog No. AAJ63394AK



Description

Description

Applications
Sodium n-dodecyl sulfate 20% aq. solution is used as an anionic surfactant. It is also used as fat emulsifier, a wetting agent, a detergent in cosmetics, pharmaceuticals and toothpastes. It also serves as a research tool in protein biochemistry. It is mainly used in detergents for laundry with many cleaning applications.

Solubility
Miscible with ethanol.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

Sodium n-dodecyl sulfate (SDS)
20% aq. soln.
CH3(CH2)11OSO3Na
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium;dodecyl sulfate
3423265
288.38
Colorless
151-21-3
C12H25NaO4S
MFCD00036175
CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
288.378
CHEBI:8984
Liquid
250mL

Chemical Identifiers

151-21-3
288.378
DBMJMQXJHONAFJ-UHFFFAOYSA-M
CHEBI:8984
CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
C12H25NaO4S
MFCD00036175
3423265
sodium;dodecyl sulfate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents