Learn More
Thermo Scientific Chemicals Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade
Supplier: Thermo Scientific Chemicals J6760636
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium n-dodecyl sulfate | |
∼206°C | |
C12H25NaO4S | |
500 g | |
3599286 | |
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium;dodecyl sulfate | |
3423265 | |
288.38 | |
Molecular Biology |
151-21-3 | |
White | |
MFCD00036175 | |
UN2926 | |
14,8636 | |
Soluble in water | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
CHEBI:8984 | |
99% | |
Solid |
Chemical Identifiers
151-21-3 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
3423265 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
C12H25NaO4S | |
MFCD00036175 | |
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal | |
CHEBI:8984 |
Safety and Handling
GHS H Statement
H228-H311-H302-H315-H319-H335
Flammable solid.
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302+H332-H315-H318-H335-H500
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 205-788-1
RTECSNumber : WT1050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only