missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Myristate 98.0+%, TCI America™

Manufacturer:  TCI America M048325G

 View more versions of this product

Catalog No. M048325G



Specifications

Specifications

Sodium Myristate
C14H27NaO2
Myristic Acid Sodium Salt
CCCCCCCCCCCCCC(=O)[O-].[Na+]
250.358
250.36
≥98.0% (GC,T)
25g
822-12-8
MFCD00042653
JUQGWKYSEXPRGL-UHFFFAOYSA-M
sodium;tetradecanoate
23693095
Crystal-Powder at 20°C
White

Chemical Identifiers

822-12-8
250.358
JUQGWKYSEXPRGL-UHFFFAOYSA-M
23693095
CCCCCCCCCCCCCC(=O)[O-].[Na+]
C14H27NaO2
MFCD00042653
Myristic Acid Sodium Salt
sodium;tetradecanoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0611&(7)-0973

RTECSNumber : QH4455000

TSCA : Yes

Documents