Learn More
Sodium molybdenum oxide, anhydrous, Mo 46.2%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01220922
Description
It is used as an analytical reagent, corrosion inhibitor, and non-oxidizing anodic inhibitor. It reduces the nitrite requirement of fluids inhibited with nitrite-amine and improves the corrosion protection of carboxylate salt fluids. It is utilized in pigments, dyes, anti-freezing agent, cutting oils, metal finishing agent, brightening agent for zinc, additive for fertilizers and micronutrients.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium molybdenum oxide | |
Anhydrous | |
White | |
100 g | |
Na2MoO4 | |
14,8645 | |
It is soluble in water. | |
[O-][Mo](=O)(=O)[O-].[Na+].[Na+] | |
205.926 | |
CHEBI:75215 | |
Mo 46.2% | |
1.714 | |
3.72 g/mL |
687°C | |
7631-95-0 | |
Powder | |
MoNa2O4 | |
MFCD00003486 | |
sodium molybdate, disodium molybdate, sodium molybdate vi, natriummolybdat, molybdic acid, disodium salt, natriummolybdat german, sodium molybdate van, sodium molybdate na2moo4, unii-948qaq08i1, ccris 5442 | |
TVXXNOYZHKPKGW-UHFFFAOYSA-N | |
disodium;dioxido(dioxo)molybdenum | |
61424 | |
205.92 | |
Odorless | |
Hygroscopic |
Chemical Identifiers
7631-95-0 | |
205.926 | |
TVXXNOYZHKPKGW-UHFFFAOYSA-N | |
61424 | |
disodium;dioxido(dioxo)molybdenum |
MoNa2O4 | |
MFCD00003486 | |
sodium molybdate, disodium molybdate, sodium molybdate vi, natriummolybdat, molybdic acid, disodium salt, natriummolybdat german, sodium molybdate van, sodium molybdate na2moo4, unii-948qaq08i1, ccris 5442 | |
CHEBI:75215 | |
[O-][Mo](=O)(=O)[O-].[Na+].[Na+] |
Safety and Handling
GHS H Statement
H332
Harmful if inhaled.
P260-P271-P304+P340-P312-P314-P501c
H335-H373
EINECSNumber : 231-551-7
RTECSNumber : QA5085000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only