missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium molybdate(VI) dihydrate, ACS reagent, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC446365000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Sodium molybdate(VI) dihydrate | |
500g | |
Na2MoO4·2H2O | |
15,8776 | |
Solubility in water: 560g/L (o°C). Other solubilities: 1150g/l (100°C) | |
[Na+].[Na+].[O-][Mo]([O-])(=O)=O | |
205.93 | |
CHEBI:75213 | |
ACS Reagent | |
0.001% max | |
0.0005% max | |
3.28 | |
0.005% max |
10102-40-6 | |
MoNa2O4 | |
MFCD00149170 | |
dipotassium dihydrate molybdate, disodium dihydrate molybdate, disodium molybdate dihydrate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, na2moo4.2h2o, sodium molybdate 2 hydrate, sodium molybdate dihydrate, sodium molybdate vi dihydrate, unii-8f2sxi1704 | |
TVXXNOYZHKPKGW-UHFFFAOYSA-N | |
disodium;dioxido(dioxo)molybdenum;dihydrate | |
16211258 | |
241.95 | |
0.005% max. (in water) | |
Plastic bottle | |
Solubility in water: 560g/L (o°C). Other solubilities: 1150g/l (100°C) | |
0.015% max | |
3.2800g/mL |
Chemical Identifiers
10102-40-6 | |
205.93 | |
TVXXNOYZHKPKGW-UHFFFAOYSA-N | |
16211258 | |
disodium;dioxido(dioxo)molybdenum;dihydrate |
MoNa2O4 | |
MFCD00149170 | |
dipotassium dihydrate molybdate, disodium dihydrate molybdate, disodium molybdate dihydrate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, na2moo4.2h2o, sodium molybdate 2 hydrate, sodium molybdate dihydrate, sodium molybdate vi dihydrate, unii-8f2sxi1704 | |
CHEBI:75213 | |
[Na+].[Na+].[O-][Mo]([O-])(=O)=O |
RUO – Research Use Only