missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium molybdate(VI) dihydrate, ACS reagent, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC446361000

 View more versions of this product

Catalog No. AC446361000



Description

Description

Specifications

Specifications

Sodium molybdate(VI) dihydrate
MoNa2O4·2H2O
0.001% max
dipotassium dihydrate molybdate, disodium dihydrate molybdate, disodium molybdate dihydrate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, na2moo4.2h2o, sodium molybdate 2 hydrate, sodium molybdate dihydrate, sodium molybdate vi dihydrate, unii-8f2sxi1704
FDEIWTXVNPKYDL-UHFFFAOYSA-N
disodium;dioxido(dioxo)molybdenum;dihydrate
16211258
241.95
0.005% max
0.005% max. (in water)
0.0005% max
3.28
100g
10102-40-6
Na2MoO4·2H2O
15,8776
Solubility in water: 560g/L (o°C). Other solubilities: 1150g/l (100°C)
O.O.[O-][Mo](=O)(=O)[O-].[Na+].[Na+]
241.95
CHEBI:75213
ACS Reagent
3.2800g/mL
Plastic bottle
Solubility in water: 560g/L (o°C). Other solubilities: 1150g/l (100°C)
0.015% max

Chemical Identifiers

10102-40-6
241.95
dipotassium dihydrate molybdate, disodium dihydrate molybdate, disodium molybdate dihydrate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, na2moo4.2h2o, sodium molybdate 2 hydrate, sodium molybdate dihydrate, sodium molybdate vi dihydrate, unii-8f2sxi1704
CHEBI:75213
O.O.[O-][Mo](=O)(=O)[O-].[Na+].[Na+]
MoNa2O4·2H2O
FDEIWTXVNPKYDL-UHFFFAOYSA-N
16211258
disodium;dioxido(dioxo)molybdenum;dihydrate
SDS Documents