Please login to your online account to display your discounted pricing

Sodium Molybdate, Dihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$525.40 - $1,891.66

Chemical Identifiers

Chemical Name or Material Sodium Molybdate
CAS 10102-40-6
Name Note Dihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na2MoO4·2H2O
Formula Weight 241.95
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004177

 
avantor performance
376401
Poly Bottle 500g Each for $525.40

02004176

 
avantor performance
376405
Poly Bottle 2.5kg Each for $1,891.66
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 241.95
Chemical Name or Material Sodium Molybdate
Name Note Dihydrate
Packaging Poly Bottle
Molecular Formula Na2MoO4·2H2O
CAS 10102-40-6
Quantity 500g
Specific Gravity 1L = 3.28kg