Please login to your online account to display your discounted pricing

Sodium Molybdate ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sodium Molybdate
Name Note Dihydrate
CAS 10102-40-6
Purity Grade ACS Grade
Molecular Formula Na2MoO4·2H2O
Formula Weight 241.95
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK7782212

 
avantor performance
778206
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 241.95
Quantity 2.5kg
Chemical Name or Material Sodium Molybdate
Name Note Dihydrate
Packaging Poly
Molecular Formula Na2MoO4·2H2O
CAS 10102-40-6