Please login to your online account to display your discounted pricing

Sodium Molybdate ACS AR, Macron Fine Chemicals™


macron

Quantity 2.5kg
Packaging Poly

Manufacturer: avantor performance  778206

 View more versions of this product

Catalog No. MK7782212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Grade
Formula Weight 241.95
Quantity 2.5kg
Chemical Name or Material Sodium Molybdate
Name Note Dihydrate
Packaging Poly
Molecular Formula Na2MoO4·2H2O
CAS 10102-40-6

Specifications

Chemical Name or Material Sodium Molybdate
Name Note Dihydrate
CAS 10102-40-6
Purity Grade ACS Grade
Molecular Formula Na2MoO4·2H2O
Formula Weight 241.95