missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Methyl Sulfate 98.0+%, TCI America™

Manufacturer:  TCI America M0433250G

 View more versions of this product

Catalog No. M0433250G



Specifications

Specifications

Sodium Methyl Sulfate
250g
CH3NaO4S
sodium methyl sulfate, methylsulfuric acid sodium salt, methyl sodium sulfate, sodium methyl sulphate, sodium monomethylsulfate, unii-38w76yj3jn, sodium methylsulfate, methyl sulfate sodium salt, methyl sulfate, sodium salt, sulfuric acid, monomethyl ester, sodium salt
COS(=O)(=O)[O-].[Na+]
134.081
134.08
≥98.0% (T)
White
512-42-5
MFCD00013457
DZXBHDRHRFLQCJ-UHFFFAOYSA-M
sodium;methyl sulfate
2735086
Crystal-Powder at 20°C

Chemical Identifiers

512-42-5
134.081
DZXBHDRHRFLQCJ-UHFFFAOYSA-M
2735086
COS(=O)(=O)[O-].[Na+]
CH3NaO4S
MFCD00013457
sodium methyl sulfate, methylsulfuric acid sodium salt, methyl sodium sulfate, sodium methyl sulphate, sodium monomethylsulfate, unii-38w76yj3jn, sodium methylsulfate, methyl sulfate sodium salt, methyl sulfate, sodium salt, sulfuric acid, monomethyl ester, sodium salt
sodium;methyl sulfate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-4121

TSCA : Yes

Documents