missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium methoxide, ACS reagent, 0.5M solution in methanol, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445881000

 View more versions of this product

Catalog No. AC445881000



Description

Description

Specifications

Specifications

Sodium methoxide
124-41-4,67-56-1
CH3ONa
15,8775
Solubility in water: reacts
C[O-].[Na+]
54.02
54.02
Passes test
AcroSeal™ Glass bottle
0.8
-98.0°C
11°C
0.5M solution in methanol, AcroSeal™
CH3NaO
MFCD00012179
feldalat nm, hsdb 755, methanol, sodium salt, methoxysodium, methylate de sodium french, metilato sodico spanish, sodium methanolate, sodium methoxide, sodium methylate, unii-ig663u5emc
WQDUMFSSJAZKTM-UHFFFAOYSA-N
sodium;methanolate
10942334
ACS Reagent
0.8000g/mL
Solubility in water: reacts.
65.0°C
100mL

Chemical Identifiers

124-41-4
54.02
WQDUMFSSJAZKTM-UHFFFAOYSA-N
10942334
C[O-].[Na+]
CH3NaO
MFCD00012179
feldalat nm, hsdb 755, methanol, sodium salt, methoxysodium, methylate de sodium french, metilato sodico spanish, sodium methanolate, sodium methoxide, sodium methylate, unii-ig663u5emc
sodium;methanolate
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact

GHS Signal Word: Danger

EINECSNumber : 204-699-5

SDS Documents