Please login to your online account to display your discounted pricing

Sodium methoxide, 5.4M (30 wt.%) solution in methanol, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC428361000

 View more versions of this product

Catalog No. AC428361000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 54.02
Quantity 100mL
Packaging AcroSeal™ Glass Bottle
Density 0.9700g/mL
Merck Index 15, 8775
Linear Formula CH3ONa
CAS 67-56-1
Chemical Name or Material Sodium methoxide
Molecular Formula CH3NaO
MDL Number MFCD00012179
CAS 124-41-4
Synonym Sodium methylate
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. Causes damage to organs. Flammable liquid and vapor. Reacts violently with water.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Immediately call a POISON CENTER or doctor/physician.

Specifications

Chemical Name or Material Sodium methoxide
CAS 124-41-4
CAS 67-56-1
Molecular Formula CH3NaO
Linear Formula CH3ONa
Formula Weight 54.02
MDL Number MFCD00012179
Synonym Sodium methylate
Accessories

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Immediately call a POISON CENTER or doctor/physician.

GHS P Statement Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. Causes damage to organs. Flammable liquid and vapor. Reacts violently with water.