missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium methoxide, ≈25% w/w in methanol

Manufacturer:  Alfa Aesar™ 46585AP

 View more versions of this product

Catalog No. AA46585AP



Specifications

Specifications

Sodium methoxide
29°C (84°F)
92°C
-2°C
124-41-4
NaOCH3
UN1289
14,8643
≈ 25% w/w in methanol
C[O-].[Na+]
54.024
54.02
0.945g/mL
Air & Moisture Sensitive
Colorless to Yellow
500mL
CH3NaO
MFCD00012179
3592982
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
WQDUMFSSJAZKTM-UHFFFAOYSA-N
sodium;methanolate
10942334
Liquid

Chemical Identifiers

124-41-4
54.024
WQDUMFSSJAZKTM-UHFFFAOYSA-N
10942334
C[O-].[Na+]
CH3NaO
MFCD00012179
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
sodium;methanolate
Safety and Handling

Safety and Handling

GHS H Statement
H311-H331-H370-H314-H226
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: SODIUM METHYLATE SOLUTIONS

RTECSNumber : PC3570000

TSCA : TSCA

SDS Documents