Learn More
Sodium methoxide, 0.5M solution in methanol, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 427228000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
65°C | |
-98°C | |
Colorless | |
800 mL | |
CH3ONa | |
11°C | |
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 | |
WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
sodium;methanolate | |
10942334 | |
0.45 to 0.60M | |
0.8 |
Sodium methoxide | |
124-41-4,67-56-1 | |
Liquid | |
CH3NaO | |
MFCD00012179 | |
15, 8775 | |
Solubility in water: reacts | |
C[O-].[Na+] | |
54.02 | |
54.02 | |
AcroSeal™ Glass Bottle | |
0.8000g/mL |
Chemical Identifiers
124-41-4 | |
54.02 | |
WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
10942334 | |
C[O-].[Na+] |
CH3NaO | |
MFCD00012179 | |
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 | |
sodium;methanolate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact
GHS Signal Word: Danger
EINECSNumber : 204-699-5
RUO – Research Use Only