Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium methacrylate, 95%

$69.58 - $240.12

Chemical Identifiers

Chemical Name or Material Sodium methacrylate
CAS 5536-61-8
Molecular Formula C4H5NaO2
Formula Weight 108.07
MDL Number MFCD00045886
Beilstein 3567418
Synonym Methacrylic acid sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0927218

  SDS
alfa aesar™
L0927218
50g Each for $69.58

AAL0927230

  SDS
alfa aesar™
L0927230
250g Each for $240.12
Description & Specifications

Specifications

Formula Weight 108.07
Chemical Name or Material Sodium methacrylate
TSCA TSCA
Assay 95%
Molecular Formula C4H5NaO2
MDL Number MFCD00045886
CAS 5536-61-8
Beilstein 3567418
Synonym Methacrylic acid sodium salt
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 50g

Applications
Used in thermoplastic acrylic resin and dispersants.

Solubility
Soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.