Learn More
Sodium metavanadate, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 269210010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Sodium metavanadate | |
13718-26-8 | |
96% | |
NaVO3 | |
15,8774 | |
Solubility in water: soluble | |
[O-][V](=O)=O.[Na+] | |
121.93 | |
CHEBI:75221 | |
96% |
96% | |
1kg | |
NaO3V | |
MFCD00011125 | |
sodium metavanadate, sodium vanadate v, vanadic acid, monosodium salt, sodium vanadate meta, sodium oxido dioxo vanadium, sodium trioxovanadate 1-, metawanadan sodowy polish, vanadate vo31-, sodium, monosodium trioxovanadate 1-, ccris 6881 | |
CMZUMMUJMWNLFH-UHFFFAOYSA-N | |
sodium;oxido(dioxo)vanadium | |
4148882 | |
121.93 | |
Glass bottle |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Suspected of damaging fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Toxic to aqua
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Call a PO
GHS Signal Word: Danger
EINECSNumber : 237-272-7
RTECSNumber : YW1050000
TSCA : TSCA
RUO â Research Use Only