Sodium metatungstate monohydrate, Alfa Aesar™
Manufacturer:  Alfa Aesar 2253456
 View more versions of this product
Catalog No. AA2253456
Specifications
Specifications
Sodium metatungstate monohydrate
Na6O39W12·H2O
White powder
2986.13 (2968.12 Anhydrous)
Odorless
4 x 250g
12141-67-2
MFCD00061466
Sodium tungstate (meta), Sodium polytungstate
Powder
White
Chemical Identifiers
12141-67-2
Na6O39W12·H2O
MFCD00061466
Sodium tungstate (meta), Sodium polytungstate
Safety and Handling
Safety and Handling
H318-H302-H402-H412: Causes serious eye damage. Harmful if swallowed. Harmful to aquatic life. Harmful to aquatic life with long lasting effects.
SDS Documents