Learn More
Sodium metatungstate, 2.82 density solution in water, stabilized, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 304100500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Sodium metatungstate | |
12141-67-2 | |
Na6[H2W12O40] | |
2.8200g/mL | |
2986.13 | |
Glass bottle | |
50mL |
2.82 Density Solution in Water, Stabilized | |
H2Na6O40W12 | |
MFCD00150663 | |
Sodium polytungstate | |
≥67% (WO3) | |
2.82 |
Chemical Identifiers
12141-67-2 | |
H2Na6O40W12 | |
MFCD00150663 | |
Sodium polytungstate |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
TSCA : TSCA
RUO â Research Use Only