missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium metasilicate pentahydrate, tech., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 014103A1

 View more versions of this product

Catalog No. AA14103A1



Description

Description

Sodium metasilicate pentahydrate is used as a starting material in the preparation of silicas and zeolites. It finds application in detergents and cleaners, pulp and paper production. It is also used in soil stabilization, sealing, adhesives and binders. Further, it is used in titanium dioxide production, paints for masonry and glass, spray coatings for tunnel construction. In addition to this, it is used in fruit and vegetable washes and sanitizers for food-contact surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium metasilicate pentahydrate
10213-79-3
Granules
H10Na2O8Si
UN3253
sodium metasilicate pentahydrate, silicic acid, disodium salt, pentahydrate, disodium oxosilanediolate pentahydrate, silicic acid h2sio3 , disodium salt, pentahydrate, 2na.so3i.5h2o, sodium meta-silicate pentahydrate, sodium silicate solution water glass, disodium dioxido oxo silane pentahydrate, sodium metasilicate pentahydrate, tech., disodium oxosilanebis olate pentahydrate
ADPGKKZKGXANON-UHFFFAOYSA-N
disodium;dioxido(oxo)silane;pentahydrate
61495
Technical
1.75 g/mL
72°C
White
1 kg
MFCD00149176
14,8642
Soluble in cold water.
O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]
212.137
212.14 (122.06 Anhydrous)
Odorless

Chemical Identifiers

10213-79-3
212.137
ADPGKKZKGXANON-UHFFFAOYSA-N
61495
O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]
H10Na2O8Si
MFCD00149176
sodium metasilicate pentahydrate, silicic acid, disodium salt, pentahydrate, disodium oxosilanediolate pentahydrate, silicic acid h2sio3 , disodium salt, pentahydrate, 2na.so3i.5h2o, sodium meta-silicate pentahydrate, sodium silicate solution water glass, disodium dioxido oxo silane pentahydrate, sodium metasilicate pentahydrate, tech., disodium oxosilanebis olate pentahydrate
disodium;dioxido(oxo)silane;pentahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: DISODIUM TRIOXOSILICATE

EINECSNumber : 229-912-9

RTECSNumber : VV9287500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only