missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium metasilicate, anhydrous, tech., Alfa Aesar™

Manufacturer:  Alfa Aesar 14106A1

 View more versions of this product

Catalog No. AA14106A1



Specifications

Specifications

Sodium metasilicate
Na2O3Si
White granules
NTHWMYGWWRZVTN-UHFFFAOYSA-N
disodium;dioxido(oxo)silane
23266
122.06
Technical
Odorless
1kg
6834-92-0
MFCD00003492
sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
[O-][Si](=O)[O-].[Na+].[Na+]
122.062
CHEBI:60720
Granules
2.61g/cm3
White

Chemical Identifiers

6834-92-0
122.062
NTHWMYGWWRZVTN-UHFFFAOYSA-N
23266
disodium;dioxido(oxo)silane
Na2O3Si
MFCD00003492
sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
CHEBI:60720
[O-][Si](=O)[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

H314-H318-H302-H335: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. May cause respiratory irritation.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 229-912-9

Documents