Learn More
Sodium metaperiodate, 98%, Thermo Scientific Chemicals
Source of Periodic Acid, As An Analytical Reagent, and As An Oxidizing Agent
Supplier: Thermo Scientific Chemicals 01379836
Description
Source of periodic acid, as an analytical reagent, and as an oxidizing agent Sodium metaperiodate is used as an oxidizing reagent involved in the oxidation of cellulose. It is involved in the cleavage of vicinal diols to prepare two aldehydes. This oxidation is commonly used in selectively label RNA, saccharides and other compounds with vicinal diols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium metaperiodate | |
7790-28-5 | |
Powder | |
98% | |
MFCD00003534 | |
Soluble in water,sulfuric acid,nitric acid and acetic acid. | |
[Na+].[O-][I](=O)(=O)=O | |
213.89 | |
CHEBI:75226 | |
Odorless | |
Colorless powder |
300°C (decomposition) | |
Colorless | |
500 g | |
INaO4 | |
sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
JQWHASGSAFIOCM-UHFFFAOYSA-M | |
sodium;periodate | |
23667635 | |
213.89 | |
Hygroscopic; Light sensitive | |
4.174 g/cm3 |
Chemical Identifiers
7790-28-5 | |
213.89 | |
JQWHASGSAFIOCM-UHFFFAOYSA-M | |
23667635 | |
sodium;periodate |
INaO4 | |
MFCD00003534 | |
sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
CHEBI:75226 | |
[Na+].[O-][I](=O)(=O)=O |
Safety and Handling
P210-P220-P221-P260-P264b-P270-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P501c
H271-H314-H335-H372
DOTInformation : Hazard Class: 5.1; Packaging Group: I
EINECSNumber : 232-197-6
RTECSNumber : SD4550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only