Please login to your online account to display your discounted pricing

Sodium metaborate tetrahydrate, 98.5%, extra pure, ACROS Organics™


Quantity 10kg
Packaging Plastic drum

Manufacturer: thermo fisher scientific  AC211630100

 View more versions of this product

Catalog No. AC211630100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Extra Pure
Physical Form Crystals or Crystalline Powder
Assay ≥98.0%
Formula Weight 137.86
Quantity 10kg
Color White
Chemical Name or Material Sodium metaborate tetrahydrate
Name Note p.a.
Packaging Plastic drum
Assay Percent Range 98.5%
Molecular Formula BNaO2·4H2O
Linear Formula NaBO2·4H2O
MDL Number MFCD00149205
CAS 10555-76-7
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility. Suspected of damaging the unborn child.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Obtain special instructions before use.
IF exposed or concerned: Get medical attention/advice.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Sodium metaborate tetrahydrate
Name Note p.a.
CAS 10555-76-7
Purity Grade Extra Pure
Assay ≥98.0%
Assay Percent Range 98.5%
Molecular Formula BNaO2·4H2O
Linear Formula NaBO2·4H2O
Formula Weight 137.86
MDL Number MFCD00149205
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Obtain special instructions before use.
IF exposed or concerned: Get medical attention/advice.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility. Suspected of damaging the unborn child.