Please login to your online account to display your discounted pricing

Sodium meta-Silicate Nonahydrate (Crystalline/Certified), Fisher Chemical


Quantity 3kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  S4083

 View more versions of this product

Catalog No. S408-3

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Formula Weight 284.2
Melting Point 72°C
Quantity 3kg
DOT Information DOT Class 8, : Corrosive
Physical Form Solid
Molecular Formula Na2SiO3 · 9 H2O
Viscosity [NULL]
Color White
pH 12.4
Chemical Name or Material Sodium meta-Silicate Nonahydrate
Heavy Metals (as Pb) 0.001% max.
Identification Pass Test
Iron (Fe) 0.005% max.
Packaging Poly Bottle
Sulfate 0.01% max.
MDL Number MFCD00149175
CAS 13517-24-3
ChemAlert Storage Symbol White
Synonym Sodium Silicate
Chloride 0.02% max.

Specifications

Chemical Name or Material Sodium meta-Silicate Nonahydrate
CAS 13517-24-3
Purity Grade Certified Grade
Physical Form Solid
Molecular Formula Na2SiO3 · 9 H2O
Formula Weight 284.2
MDL Number MFCD00149175
Synonym Sodium Silicate
Accessories
 

SDS & Safety Documents