missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium meta-Silicate Nonahydrate (Crystalline/Certified), Fisher Chemical™

$311.38 - $311.38

Chemical Identifiers

CAS 13517-24-3
Molecular Formula H18Na2O12Si
Molecular Weight (g/mol) 284.197
MDL Number MFCD00149175
InChI Key PHIQPXBZDGYJOG-UHFFFAOYSA-N
Synonym 2na.so3i.9h2o, disodium dioxido oxo silane nonahydrate, disodium oxosilanebis olate nonahydrate, disodium oxosilanediolate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, sodium meta-silicate nonahydrate, sodium metasilicate nonahydrate, sodium oxosilanebis olate-water 1/9, sodium silicate nonahydrate, unii-d8d44215lz
PubChem CID 61639
IUPAC Name disodium;dioxido(oxo)silane;nonahydrate
SMILES O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
S408-500
SDS View Documents
Fisher Chemical
S408500
500g Poly Bottle
Each for $311.38
Case of 6 Each for $1,868.28
 
Description

Description

Specifications

Chemical Identifiers

13517-24-3
284.197
PHIQPXBZDGYJOG-UHFFFAOYSA-N
61639
O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]
H18Na2O12Si
MFCD00149175
2na.so3i.9h2o, disodium dioxido oxo silane nonahydrate, disodium oxosilanebis olate nonahydrate, disodium oxosilanediolate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, sodium meta-silicate nonahydrate, sodium metasilicate nonahydrate, sodium oxosilanebis olate-water 1/9, sodium silicate nonahydrate, unii-d8d44215lz
disodium;dioxido(oxo)silane;nonahydrate

Specifications

13517-24-3
MFCD00149175
PHIQPXBZDGYJOG-UHFFFAOYSA-N
disodium;dioxido(oxo)silane;nonahydrate
61639
Solid
0.02% max.
Pass Test
Poly Bottle
White
12.4
Sodium meta-Silicate Nonahydrate
H18Na2O12Si
2na.so3i.9h2o, disodium dioxido oxo silane nonahydrate, disodium oxosilanebis olate nonahydrate, disodium oxosilanediolate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, sodium meta-silicate nonahydrate, sodium metasilicate nonahydrate, sodium oxosilanebis olate-water 1/9, sodium silicate nonahydrate, unii-d8d44215lz
O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]
284.197
284.2
Certified
0.001% max.
0.005% max.
0.01% max.
72°C
500g
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS
Product Certifications