Please login to your online account to display your discounted pricing

Sodium Meta-Silicate, 9-Hydrate, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  386805

 View more versions of this product

Catalog No. 02004174

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 284.2
Chemical Name or Material Sodium Meta-Silicate
Name Note 9-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Na2SiO3·9H2O
CAS 13517-24-3
Packaging Poly Bottle
Quantity 2.5kg

Specifications

Chemical Name or Material Sodium Meta-Silicate
CAS 13517-24-3
Name Note 9-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Na2SiO3·9H2O
Formula Weight 284.2